2-Chlor-3,5-dimethylpyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS35633G1 AS35633G5
ACTIAS35633G1EA 0 CHF
ACTIAS35633G1 ACTIAS35633G5
2-Chlor-3,5-dimethylpyrazin
2-Chloro-3,5-dimethylpyrazine
Formel: C₆H₇ClN₂
MW: 142,5900 g/mol
MDL Number: MFCD00126945
CAS-Nummer: 38557-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR