2-Methyl-3-(methylthio)pyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS24438G100 AS24438G25 AS24438G5
ACTIAS24438G100EA 0 CHF
ACTIAS24438G100 ACTIAS24438G25 ACTIAS24438G5
2-Methyl-3-(methylthio)pyrazin
2-Methyl-3-(methylthio)pyrazine
Formel: C₆H₈N₂S
MDL Number: MFCD00014610
CAS-Nummer: 2882-20-4
EINECS: 220-736-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR