2-Amino-6-methylpyrazin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
6-Methylpyrazin-2-amine
AS31397G1
AS31397G5
ACTIAS31397G1EA
0
CHF
ACTIAS31397G1
ACTIAS31397G5
2-Amino-6-methylpyrazin
2-Amino-6-methylpyrazine
Formel:
C₅H₇N₃ MW: 109,13 g/mol |
MDL Number:
MFCD08235193 CAS-Nummer: 5521-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...