2-Amino-3-chlor-5-methylpyrazin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Chloro-5-methylpyrazin-2-amine
AS30533G1
AS30533G5
ACTIAS30533G1EA
0
CHF
ACTIAS30533G1
ACTIAS30533G5
2-Amino-3-chlor-5-methylpyrazin
2-Amino-3-chloro-5-methylpyrazine
Formel:
C₅H₆ClN₃ MW: 143,5700 g/mol |
MDL Number:
MFCD09834805 CAS-Nummer: 89182-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...