2-Chlor-6-methylpyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS40221G1 AS40221G5
ACTIAS40221G1EA 0 CHF
ACTIAS40221G1 ACTIAS40221G5
2-Chlor-6-methylpyrazin
2-Chloro-6-methylpyrazine
Formel: C₅H₅ClN₂
MW: 128,5596 g/mol
MDL Number: MFCD00055032
CAS-Nummer: 38557-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR