2-Chlor-3-methylpyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS31734G1 AS31734G5
ACTIAS31734G1EA 0 CHF
ACTIAS31734G1 ACTIAS31734G5
2-Chlor-3-methylpyrazin
2-Chloro-3-methylpyrazine
Formel: C₅H₅ClN₂
MW: 128,56 g/mol
MDL Number: MFCD00023279
CAS-Nummer: 95-58-9
EINECS: 202-434-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR