2-Amino-6-methoxypyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS30748G1 AS30748G5
ACTIAS30748G1EA 0 CHF
ACTIAS30748G1 ACTIAS30748G5
2-Amino-6-methoxypyrazin
2-Amino-6-methoxypyrazine
Formel: C₅H₇N₃O
MW: 125,13 g/mol
MDL Number: MFCD09909689
CAS-Nummer: 6905-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR