2-Amino-5-methoxypyrazin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Methoxypyrazin-2-amine

AS30713G1 AS30713G5
ACTIAS30713G1EA 0 CHF
ACTIAS30713G1 ACTIAS30713G5
2-Amino-5-methoxypyrazin
2-Amino-5-methoxypyrazine
Formel: C₅H₇N₃O
MW: 125,13 g/mol
MDL Number: MFCD09909647
CAS-Nummer: 54013-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR