2-Amino-6-cyanpyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS31042G1 AS31042G5
ACTIAS31042G1EA 0 CHF
ACTIAS31042G1 ACTIAS31042G5
2-Amino-6-cyanpyrazin
2-Amino-6-cyanopyrazine
Formel: C₅H₄N₄
MW: 120,1100 g/mol
MDL Number: MFCD10697805
CAS-Nummer: 59489-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR