2-Chlor-3-methoxypyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS31278G1 AS31278G5
ACTIAS31278G1EA 0 CHF
ACTIAS31278G1 ACTIAS31278G5
2-Chlor-3-methoxypyrazin
2-Chloro-3-methoxypyrazine
Formel: C₅H₅ClN₂O
MW: 144.5600 g/mol
MDL Number: MFCD02684324
CAS-Nummer: 40155-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR