2-Chlor-6-(methylamino)pyrazin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 6-Chloro-N-methylpyrazin-2-amine

AS32734G1 AS32734G5
ACTIAS32734G1EA 0 CHF
ACTIAS32734G1 ACTIAS32734G5
2-Chlor-6-(methylamino)pyrazin
2-Chloro-6-(methylamino)pyrazine
Formel: C₅H₆ClN₃
MW: 143,5700 g/mol
MDL Number: MFCD09878383
CAS-Nummer: 848366-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR