2-Amino-6-chlorpyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS50153G100 AS50153G5
ACTIAS50153G100EA 0 CHF
ACTIAS50153G100 ACTIAS50153G5
2-Amino-6-chlorpyrazin
2-Amino-6-chloropyrazine
Formel: C₄H₄ClN₃
MDL Number: MFCD00055024
CAS-Nummer: 33332-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR