2-Amino-5-chlorpyrazin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-amino-5-chloropyrazine

AS1895G1 AS1895G5
ACTIAS1895G1EA 0 CHF
ACTIAS1895G1 ACTIAS1895G5
2-Amino-5-chlorpyrazin
2-Amino-5-chloropyrazine
Formel: C₄H₄ClN₃
MDL Number: MFCD03423596
CAS-Nummer: 33332-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR