2-Amino-3,5-dichlorpyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS1857G1 AS1857G5
ACTIAS1857G1EA 0 CHF
ACTIAS1857G1 ACTIAS1857G5
2-Amino-3,5-dichlorpyrazin
2-Amino-3,5-dichloropyrazine
Formel: C₄H₃Cl₂N₃
MW: 163.9900 g/mol
MDL Number: MFCD11040179
CAS-Nummer: 873-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR