2,6-Dichlor-3-cyanpyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS32047G1 AS32047G5
ACTIAS32047G1EA 0 CHF
ACTIAS32047G1 ACTIAS32047G5
2,6-Dichlor-3-cyanpyrazin
2,6-Dichlor-3-cyanpyrazin
Formel: C₅HCl₂N₃
MDL Number: MFCD13193460
CAS-Nummer: 313339-92-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR