2-Brom-5-hydroxypyrazin
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Bromopyrazin-2-ol
AS50755G1
AS50755G5
ACTIAS50755G1EA
0
CHF
ACTIAS50755G1
ACTIAS50755G5
2-Brom-5-hydroxypyrazin
2-Bromo-5-hydroxypyrazine
Formel:
C₄H₃BrN₂O |
MDL Number:
MFCD06245330 CAS-Nummer: 374063-92-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...