2-Brom-5-iodpyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS50758G1 AS50758G5
ACTIAS50758G1EA 0 CHF
ACTIAS50758G1 ACTIAS50758G5
2-Brom-5-iodpyrazin
2-Bromo-5-iodopyrazine
Formel: C₄H₂BrIN₂
MW: 284,8800 g/mol
MDL Number: MFCD08061602
CAS-Nummer: 622392-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR