2-Brom-5-chlorpyrazin

Lieferant: ACTIVATE SCIENTIFIC

AS50757G1 AS50757G5
ACTIAS50757G1EA 0 CHF
ACTIAS50757G1 ACTIAS50757G5
2-Brom-5-chlorpyrazin
2-Bromo-5-chloropyrazine
Formel: C₄H₂BrClN₂
MW: 193,4291 g/mol
MDL Number: MFCD08460074
CAS-Nummer: 912773-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR