(3S)-Tetrahydro-3-furanol

Lieferant: ACTIVATE SCIENTIFIC

AS32492G1 AS32492G5
ACTIAS32492G1EA 0 CHF
ACTIAS32492G1 ACTIAS32492G5
(3S)-Tetrahydro-3-furanol
(S)-(+)-3-Hydroxytetrahydrofuran
Formel: C₄H₈O₂
MDL Number: MFCD00064327
CAS-Nummer: 86087-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR