2,2-Dimethylcyclohexanon

Lieferant: ACTIVATE SCIENTIFIC

AS33275G1 AS33275G5
ACTIAS33275G1EA 0 CHF
ACTIAS33275G1 ACTIAS33275G5
2,2-Dimethylcyclohexanon
2,2-Dimethylcyclohexanone
Formel: C₈H₁₄O
MW: 126,1962 g/mol
MDL Number: MFCD00134382
CAS-Nummer: 1193-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR