4,4-Difluorcyclohexanon

Lieferant: ACTIVATE SCIENTIFIC

AS51104G1 AS51104G5
ACTIAS51104G1EA 0 CHF
ACTIAS51104G1 ACTIAS51104G5
4,4-Difluorcyclohexanon
4,4-Difluorocyclohexanone
Formel: C₆H₈F₂O
MW: 134,12 g/mol
MDL Number: MFCD08146635
CAS-Nummer: 22515-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR