8-Methyl-1,2,3,4-tetrahydrochinolin

Lieferant: ACTIVATE SCIENTIFIC

AS36514G1 AS36514G5
ACTIAS36514G1EA 0 CHF
ACTIAS36514G1 ACTIAS36514G5
8-Methyl-1,2,3,4-tetrahydrochinolin
8-Methyl-1,2,3,4-tetrahydrochinolin
Formel: C₁₀H₁₃N
MW: 147,2200 g/mol
MDL Number: MFCD00218100
CAS-Nummer: 52601-70-4
EINECS: 202-083-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR