2-Amino-3-fluorphenol
Lieferant: ACTIVATE SCIENTIFIC
AS34419G1
AS34419G10
AS34419G5
ACTIAS34419G1EA
0
CHF
ACTIAS34419G1
ACTIAS34419G10
ACTIAS34419G5
2-Amino-3-fluorphenol
2-Amino-3-fluorphenol
Formel:
C₆H₆FNO MW: 127,1163 g/mol |
MDL Number:
MFCD07368754 CAS-Nummer: 53981-23-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...