2-(3-Brompropoxy)tetrahydropyran
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2-(3-Bromopropoxy)tetrahydro-2H-pyran
AS23675G25
AS23675G5
ACTIAS23675G25EA
127
CHF
ACTIAS23675G25
ACTIAS23675G5
2-(3-Brompropoxy)tetrahydropyran
2-(3-Brompropoxy)tetrahydropyran
Formel:
C₈H₁₅BrO₂ MW: 223,1075 g/mol |
MDL Number:
MFCD00058593 CAS-Nummer: 33821-94-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...