2-Chlor-4,8-dimethylchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS36168G1 AS36168G5
ACTIAS36168G1EA 0 CHF
ACTIAS36168G1 ACTIAS36168G5
2-Chlor-4,8-dimethylchinolin
2-Chloro-4,8-dimethylquinoline
Formel: C₁₁H₁₀ClN
MDL Number: MFCD00760267
CAS-Nummer: 3913-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR