3-Benzo[b]thiophencarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Benzothiophene-3-carboxylic acid

AS31840G1 AS31840G5
ACTIAS31840G1EA 0 CHF
ACTIAS31840G1 ACTIAS31840G5
3-Benzo[b]thiophencarbonsäure
3-Benzo[b]thiophencarbonsäure
Formel: C₉H₆O₂S
MW: 178,22 g/mol
MDL Number: MFCD01846406
CAS-Nummer: 5381-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR