2-Benzo[b]thiophencarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

AS33483G1 AS33483G10 AS33483G5
ACTIAS33483G1EA 0 CHF
ACTIAS33483G1 ACTIAS33483G10 ACTIAS33483G5
2-Benzo[b]thiophencarbonsäure
2-Benzo[b]thiophencarbonsäure
Formel: C₉H₆O₂S
MW: 178,22 g/mol
MDL Number: MFCD00051636
CAS-Nummer: 6314-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR