2,3-Dihydrobenzo[b]furan

Lieferant: ACTIVATE SCIENTIFIC

AS21067G100 AS21067G25
ACTIAS21067G100EA 0 CHF
ACTIAS21067G100 ACTIAS21067G25
2,3-Dihydrobenzo[b]furan
2,3-Dihydrobenzo[b]furan
Formel: C₈H₈O
MDL Number: MFCD00005855
CAS-Nummer: 496-16-2
EINECS: 207-817-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR