2,3-Dihydrobenzo[b]furan-7-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,3-Dihydrobenzofuran-7-carboxylic acid

AS36604G1 AS36604G5
ACTIAS36604G1EA 0 CHF
ACTIAS36604G1 ACTIAS36604G5
2,3-Dihydrobenzo[b]furan-7-carbonsäure
2,3-Dihydrobenzo[b]furan-7-carbonsäure
Formel: C₉H₈O₃
MW: 164,1580 g/mol
MDL Number: MFCD00671526
CAS-Nummer: 35700-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR