2,3-Dihydrobenzo[b]furan-5-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,3-Dihydrobenzo[b]furan-5-carboxylicacid

AS21184G25 AS21184G5
ACTIAS21184G25EA 400 CHF
ACTIAS21184G25 ACTIAS21184G5
2,3-Dihydrobenzo[b]furan-5-carbonsäure
2,3-Dihydrobenzo[b]furan-5-carbonsäure
Formel: C₉H₈O₃
MW: 164,1580 g/mol
MDL Number: MFCD00085012
CAS-Nummer: 76429-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR