1-Ethoxymethyl-2-iodimidazol

Lieferant: ACTIVATE SCIENTIFIC

AS51150G1 AS51150G5
ACTIAS51150G1EA 0 CHF
ACTIAS51150G1 ACTIAS51150G5
1-Ethoxymethyl-2-iodimidazol
1-Ethoxymethyl-2-iodimidazol
Formel: C₆H₉IN₂O
MW: 252,05 g/mol
MDL Number: MFCD03788568
CAS-Nummer: 146697-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR