1-Methylimidazol-5-carbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Methylimidazole-5-carboxylic Acid

AS50342G1 AS50342G5
ACTIAS50342G1EA 0 CHF
ACTIAS50342G1 ACTIAS50342G5
1-Methylimidazol-5-carbonsäure
1-Methylimidazol-5-carbonsäure
Formel: C₅H₆N₂O₂
MW: 126.1133 g/mol
MDL Number: MFCD00955677
CAS-Nummer: 41806-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR