Imidazol-4-carbonsäureethylester
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Ethyl imidazole-4-carboxylate
AS31214G1
AS31214G5
ACTIAS31214G1EA
0
CHF
ACTIAS31214G1
ACTIAS31214G5
Imidazol-4-carbonsäureethylester
Imidazol-4-carbonsäureethylester
Formel:
C₆H₈N₂O₂ MW: 140.14 g/mol |
MDL Number:
MFCD07363812 CAS-Nummer: 23785-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...