Imidazol-2-carbonsäureethylester

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl imidazole-2-carboxylate, Ethyl 1H-imidazole-2-carboxylate

AS32679G1 AS32679G5
ACTIAS32679G1EA 0 CHF
ACTIAS32679G1 ACTIAS32679G5
Imidazol-2-carbonsäureethylester
Imidazol-2-carbonsäureethylester
Formel: C₆H₈N₂O₂
MW: 140,14 g/mol
MDL Number: MFCD03426031
CAS-Nummer: 33543-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR