5-Brom-2'-deoxyuridin (Broxuridin)
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU
AS35316G1
AS35316G10
AS35316G5
ACTIAS35316G1EA
0
CHF
ACTIAS35316G1
ACTIAS35316G10
ACTIAS35316G5
5-Brom-2'-deoxyuridin (Broxuridin)
5-Brom-2'-deoxyuridin (Broxuridin)
Formel:
C₉H₁₁BrN₂O₅ MW: 307,10 g/mol |
MDL Number:
MFCD00006529 CAS-Nummer: 59-14-3 EINECS: 200-415-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...