2,2-Dimethylbernsteinsäureanhydrid

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,2-Dimethylsuccinicanhydride

AS22292G100 AS22292G25
ACTIAS22292G100EA 239 CHF
ACTIAS22292G100 ACTIAS22292G25
2,2-Dimethylbernsteinsäureanhydrid
2,2-Dimethylbernsteinsäureanhydrid
Formel: C₆H₈O₃
MW: 128,1259 g/mol
MDL Number: MFCD00022608
CAS-Nummer: 17347-61-4
EINECS: 241-371-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR