4-Chlor-2-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS31348G1EA 0 CHF
ACTIAS31348G1 ACTIAS31348G5
4-Chlor-2-methoxypyridin
4-Chloro-2-methoxypyridine
Formel: C₆H₆ClNO
MW: 143,5709 g/mol
MDL Number: MFCD06738656
CAS-Nummer: 72141-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR