2-Chlor-6-methylchinolin

Lieferant: ACTIVATE SCIENTIFIC

AS36050G1 AS36050G5
ACTIAS36050G1EA 0 CHF
ACTIAS36050G1 ACTIAS36050G5
2-Chlor-6-methylchinolin
2-Chloro-6-methylquinoline
Formel: C₁₀H₈ClN
MW: 177.6302 g/mol
MDL Number: MFCD00462389
CAS-Nummer: 4295-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR