4,4,4-Trifluorbuttersäure

Lieferant: ACTIVATE SCIENTIFIC

AS34428G10 AS34428G100 AS34428G5
ACTIAS34428G10EA 0 CHF
ACTIAS34428G10 ACTIAS34428G100 ACTIAS34428G5
4,4,4-Trifluorbuttersäure
4,4,4-Trifluorbuttersäure
Formel: C₄H₅F₃O₂
MW: 142,08 g/mol
MDL Number: MFCD00077604
CAS-Nummer: 406-93-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR