2-Amino-3-chlorphenol
Lieferant: ACTIVATE SCIENTIFIC
AS36350G1
AS36350G5
ACTIAS36350G1EA
0
CHF
ACTIAS36350G1
ACTIAS36350G5
2-Amino-3-chlorphenol
2-Amino-3-chlorphenol
Formel:
C₆H₆ClNO MW: 143.5700 g/mol |
MDL Number:
MFCD11848418 CAS-Nummer: 56962-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...