5-Brom-2-methoxy-3-nitropyridin

Lieferant: ACTIVATE SCIENTIFIC

AS33486G1 AS33486G10 AS33486G5
ACTIAS33486G1EA 0 CHF
ACTIAS33486G1 ACTIAS33486G10 ACTIAS33486G5
5-Brom-2-methoxy-3-nitropyridin
5-Brom-2-methoxy-3-nitropyridin
Formel: C₆H₅BrN₂O₃
MW: 233,0200 g/mol
MDL Number: MFCD07374968
CAS-Nummer: 152684-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR