(±)-α-Bromphenylessigsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid
AS22054G100
AS22054G25
ACTIAS22054G100EA
400
CHF
ACTIAS22054G100
ACTIAS22054G25
(±)-α-Bromphenylessigsäure
(±)-α-Bromphenylessigsäure
Formel:
C₈H₇BrO₂ MW: 215,0440 g/mol |
MDL Number:
MFCD00004206 CAS-Nummer: 4870-65-9 EINECS: 225-477-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...