3,5-Dibrom-2-methoxypyridin

Lieferant: ACTIVATE SCIENTIFIC

AS31769G1 AS31769G5
ACTIAS31769G1EA 0 CHF
ACTIAS31769G1 ACTIAS31769G5
3,5-Dibrom-2-methoxypyridin
3,5-Dibromo-2-methoxypyridine
Formel: C₆H₅Br₂NO
MDL Number: MFCD05663709
CAS-Nummer: 13472-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR