2,4-Dichlor-6,7-dimethoxychinazolin

Lieferant: ACTIVATE SCIENTIFIC

AS35308G1 AS35308G10 AS35308G5
ACTIAS35308G1EA 0 CHF
ACTIAS35308G1 ACTIAS35308G10 ACTIAS35308G5
2,4-Dichlor-6,7-dimethoxychinazolin
2,4-Dichloro-6,7-dimethoxyquinazoline
Formel: C₁₀H₈Cl₂N₂O₂
MDL Number: MFCD00051733
CAS-Nummer: 27631-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR