2-(4-Morpholinyl)phenol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Morpholinophenol

AS35725G1 AS35725G5
ACTIAS35725G1EA 0 CHF
ACTIAS35725G1 ACTIAS35725G5
2-(4-Morpholinyl)phenol
2-(4-Morpholinyl)phenol
Formel: C₁₀H₁₃NO₂
MDL Number: MFCD00051674
CAS-Nummer: 41536-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR