5-Brom-2-(isopropylamino)pyrimidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: (5-Bromopyrimidin-2-yl)isopropylamine

AS51209G1 AS51209G5
ACTIAS51209G1EA 0 CHF
ACTIAS51209G1 ACTIAS51209G5
5-Brom-2-(isopropylamino)pyrimidin
5-Brom-2-(isopropylamino)pyrimidin
Formel: C₇H₁₀BrN₃
MW: 216.0784 g/mol
MDL Number: MFCD00483159
CAS-Nummer: 77476-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR