5-Brom-2-chlor-4-methoxypyrimidin

Lieferant: ACTIVATE SCIENTIFIC

AS33846G1 AS33846G10 AS33846G5
ACTIAS33846G1EA 0 CHF
ACTIAS33846G1 ACTIAS33846G10 ACTIAS33846G5
5-Brom-2-chlor-4-methoxypyrimidin
5-Bromo-2-chloro-4-methoxypyrimidine
Formel: C₅H₄BrClN₂O
MW: 223,4600 g/mol
MDL Number: MFCD03001373
CAS-Nummer: 57054-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR