6-(Methylthio)purin

Lieferant: ACTIVATE SCIENTIFIC

AS35311G1 AS35311G5
ACTIAS35311G1EA 0 CHF
ACTIAS35311G1 ACTIAS35311G5
6-(Methylthio)purin
6-(Methylthio)purine
Formel: C₆H₆N₄S
MDL Number: MFCD00005576
CAS-Nummer: 50-66-8
EINECS: 200-057-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR