1-Chlor-2,3-difluorbenzol

Lieferant: ACTIVATE SCIENTIFIC

AS34903G1 AS34903G10 AS34903G5
ACTIAS34903G1EA 0 CHF
ACTIAS34903G1 ACTIAS34903G10 ACTIAS34903G5
1-Chlor-2,3-difluorbenzol
1-Chloro-2,3-difluorobenzene
Formel: C₆H₃ClF₂
MW: 148,5400 g/mol
MDL Number: MFCD04112584
CAS-Nummer: 36556-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR