3-Brom-5-chlorphenol
Lieferant: ACTIVATE SCIENTIFIC
AS21457G25
AS21457G5
ACTIAS21457G25EA
158
CHF
ACTIAS21457G25
ACTIAS21457G5
3-Brom-5-chlorphenol
3-Brom-5-chlorphenol
Formel:
C₆H₄BrClO MW: 207,45 g/mol |
MDL Number:
MFCD07780676 CAS-Nummer: 56962-04-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...